Orders LinkContactUsLinkComponent
Multi-level removal funnels, Squibb
Multi-level removal funnels, Squibb
4000 ml, 45/40 NS
Catalog # WITG2.636.004
Supplier:   witeg Labortechnik
Multi-level removal funnels, Squibb
Catalog # WITG2.636.004
Supplier:  witeg Labortechnik
Multi-level removal funnels, Squibb
Multi-level removal funnels, Squibb
4000 ml, 45/40 NS
Price
447,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Multi-level removal funnel, Squibb, made of borosilicate glass 3.3, with PTFE-outlet valve and plastic stopper.
  • Easy to read graduation
  • Very good chemical resistance
  • Very good temperature resistance
Specifications
  • Socket size:
    45/40 NS
  • Capacity:
    4000 ml