Order Entry
Austria
Orders LinkContactUsLinkComponent
Büchner funnels

Büchner funnels

60 mm, 125 ml, 4
Catalog # WITG2.367.124
Supplier:   witeg Labortechnik
Büchner funnels
Catalog # WITG2.367.124
Supplier:  witeg Labortechnik
Büchner funnels
Büchner funnels

60 mm, 125 ml, 4

Price
51,40 €
Log in for availability
1 * 1 ST
Product Details & Documents

Funnels, made of borosilicate glass 3.3, with sintered disc, for convenient filtration of solutions.

  • No filter papers required
  • Highly resistant to most corrosive reagents including ammonia and concentrated sulphuric acid
  • Very good temperature resistance
Specifications
  • Plate Ø:
    60 mm
  • Porosity:
    4
  • Capacity:
    125 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.