Orders LinkContactUsLinkComponent
Graduated pipettes, with piston, Class B
Graduated pipettes, with piston, Class B
Supplier:   witeg Labortechnik
Graduated pipettes, with piston, Class B
Supplier:  witeg Labortechnik
Certificates
Graduated pipettes, with piston, Class B
Graduated pipettes, with piston, Class B
5 Options Available Below
Product Details & Documents
Glass, class DIN-B
  • With ISO-COLOR-Code
  • With interchangeable piston and tempered tips
  • With amber stain DIFFICO graduation and inscriptions
  • Short line graduation
  • Complete delivery
Product Family Options
Product Information
  • Capacity
    Division
    Tolerance
    Colour code
    Availability
    Price
  • 1 ml
    0,01 ml
    0,008 ± ml
    Yellow
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 15,30 €
    15,30 €
    1 * 1 ST
       15,30 €
  • 5 ml
    0,05 ml
    0,04 ± ml
    Black
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 15,60 €
    15,60 €
    1 * 1 ST
       15,60 €
  • 10 ml
    0,1 ml
    0,08 ± ml
    Brown
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 17,50 €
    17,50 €
    1 * 1 ST
       17,50 €
  • 25 ml
    0,1 ml
    0,15 ± ml
    White
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 28,20 €
    28,20 €
    1 * 1 ST
       28,20 €
  • 0,5 ml
    0,01 ml
    0,008 ± ml
    2 × yellow
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 16,70 €
    16,70 €
    1 * 1 ST
       16,70 €