Volumetric Flasks, Class A
Supplier: witeg Labortechnik
Jump to:
About this item
Borosilicate glass 3.3, with or without stopper.
- DE-M marked according to the German calibration law
- Inclusive batch certificate
Certifications: Tolerance according to DIN EN ISO 1042
17 Options Available Below
Product Details & Documents
Borosilicate glass 3.3, with or without stopper.
- DE-M marked according to the German calibration law
- Inclusive batch certificate
Certifications: Tolerance according to DIN EN ISO 1042
Product Family Options
Blue graduated
Product Information
- CapacityNSToleranceAvailabilityPrice
Brown graduated
Product Information
- CapacityNSToleranceAvailabilityPrice
Blue graduated without stopper
Product Information
- CapacityNSToleranceAvailabilityPrice
- Specifications:New ProductNew Product