Order Entry
Austria
Orders LinkContactUsLinkComponent
Pump sampler sets

Pump sampler sets

Pump sampler set with hand pump, stainless steel bottle adapter, PE 100 ml bottles, PE 10 m sampling tube, tube cutter, stainless steel hose weight, and case, Stainless steel 316L
Catalog # SAMP5020S-02
Supplier:   Sampling Systems
Pump sampler sets
Catalog # SAMP5020S-02
Supplier:  Sampling Systems
Pump sampler sets
Pump sampler sets

Pump sampler set with hand pump, stainless steel bottle adapter, PE 100 ml bottles, PE 10 m sampling tube, tube cutter, stainless steel hose weight, and case, Stainless steel 316L

Price
1 750,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

These pump samplers allow for the quick sampling of liquids.

  • Reduced contamination by using new tube and bottles for each sample
  • Only the tube and collecting bottle come into contact with the products
  • High efficiency and ability to pump water to a height of about four meters

Suitable for low to medium viscosity liquids.

Specifications
  • Description:
    Pump sampler set with hand pump, stainless steel bottle adapter, PE 100 ml bottles, PE 10 m sampling tube, tube cutter, stainless steel hose weight, and case
  • Material:
    Stainless steel 316L
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • PTFE sampling tube
    PTFE sampling tube
    SAMP5020T-100
    Log in for availability
    1 * 100 m - 779,00 €

Product Information

  • Catalog #
    Availability
    Price
  • PTFE sampling tube, 5 m
    PTFE sampling tube, 5 m
    SAMP5020T-05
    Log in for availability
    1 * 1 ST - 80,90 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.