Orders LinkContactUsLinkComponent
Chromatographic columns with discs
Chromatographic columns with discs
Supplier:   witeg Labortechnik
Chromatographic columns with discs
Supplier:  witeg Labortechnik
Certificates
Chromatographic columns with discs
Chromatographic columns with discs
4 Options Available Below
Product Details & Documents
These borosilicate glass chromatography columns have fused-in sintered glass discs, ST-stopcock, with solid plug and screw-thread retaining nut.
Product Family Options
Product Information
  • Ø int.
    Length
    Capacity
    Socket size
    Availability
    Price
  • WITG2.132.005
    Chromatography column with fused-in sintered glass disc with ST-stopcock
    Chromatography column with fused-in sintered glass disc with ST-stopcock
    20,00 mm
    200 mm
    60 ml
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 86,20 €
    86,20 €
    1 * 1 ST
       86,20 €
  • WITG2.132.008
    Chromatography column with fused-in sintered glass disc with ST-stopcock
    Chromatography column with fused-in sintered glass disc with ST-stopcock
    20,00 mm
    300 mm
    90 ml
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 76,30 €
    76,30 €
    1 * 1 ST
       76,30 €
  • WITG2.132.001
    Chromatography column with fused-in sintered glass disc with ST-stopcock
    Chromatography column with fused-in sintered glass disc with ST-stopcock
    10,00 mm
    100 mm
    8 ml
    14/23 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 71,50 €
    71,50 €
    1 * 1 ST
       71,50 €
  • WITG2.132.014
    Chromatography column with fused-in sintered glass disc with ST-stopcock
    Chromatography column with fused-in sintered glass disc with ST-stopcock
    40,00 mm
    800 mm
    1000 ml
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 118,00 €
    118,00 €
    1 * 1 ST
       118,00 €