Orders LinkContactUsLinkComponent
Adapters for distilling receivers
Adapters for distilling receivers
14/23 NS
Catalog # WITG1.920.001
Supplier:   witeg Labortechnik
Adapters for distilling receivers
Catalog # WITG1.920.001
Supplier:  witeg Labortechnik
Adapters for distilling receivers
Adapters for distilling receivers
14/23 NS
Price
51,10 €
Log in for availability
1 * 1 ST
Product Details & Documents
Borosilicate glass 3,3
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Specifications
  • Socket size:
    14/23 NS