Orders LinkContactUsLinkComponent
Buckets, Round, PP
Buckets, Round, PP
Buckets, Round, PP
Certificates
Buckets, Round, PP
20 Options Available Below
Product Details & Documents
Buckets made of PP, white.
  • With or without UN approval
  • Very good chemical resistance
  • Very good temperature resistance
Universal multipurpose buckets with handle.
Product Family Options
Product Information
  • Colour
    Capacity
    Ø
    Height
    Availability
    Price
  • RIXI7-1001-006
    Bucket with plastic handle, not UN-approved (without lid, must be ordered separately)
    Bucket with plastic handle, not UN-approved (without lid, must be ordered separately)
    White
    0,6 L
    190 mm
    102 mm
    Log in for availability
    Log in for availability
    1 * 435 ST
    1 * 435 ST - 788,00 €
    788,00 €
    1 * 435 ST
       788,00 €
  • RIXI7-1001-010
    Bucket with plastic handle and lid, not UN-approved
    Bucket with plastic handle and lid, not UN-approved
    White
    1,0 L
    131 mm
    125,5 mm
    Log in for availability
    Log in for availability
    1 * 180 ST
    1 * 180 ST - 289,00 €
    289,00 €
    1 * 180 ST
       289,00 €
  • RIXIU-7S-1001-018K
    Bucket with plastic handle and lid, not UN-approved
    Bucket with plastic handle and lid, not UN-approved
    White
    1,8 L
    168,5 mm
    128 mm
    Log in for availability
    Log in for availability
    1 * 180 ST
    1 * 180 ST - 886,00 €
    886,00 €
    1 * 180 ST
       886,00 €
  • RIXIU-7S-1001-020K
    Bucket with plastic handle, not UN-approved
    Bucket with plastic handle, not UN-approved
    White
    2,0 L
    179 mm
    132 mm
    Log in for availability
    Log in for availability
    1 * 120 ST
    1 * 120 ST - 687,00 €
    687,00 €
    1 * 120 ST
       687,00 €
  • RIXI7-1001-036-K
    Tamper proof bucket with plastic handle and lid, not UN-approved
    Tamper proof bucket with plastic handle and lid, not UN-approved
    White
    3,6 L
    192 mm
    142,5 mm
    Log in for availability
    Log in for availability
    1 * 1.080 ST
    1 * 1.080 ST - 4 900,00 €
    4 900,00 €
    1 * 1.080 ST
       4 900,00 €
  • RIXIU-7S-1001-025K
    Tamper proof bucket with plastic handle, not UN-approved (without lid, must be ordered seperately)
    Tamper proof bucket with plastic handle, not UN-approved (without lid, must be ordered seperately)
    White
    2,5 L
    179 mm
    159 mm
    Log in for availability
    Log in for availability
    1 * 120 ST
    1 * 120 ST - 519,00 €
    519,00 €
    1 * 120 ST
       519,00 €
  • RIXIU-7S-1001-030K
    Bucket with plastic handle and lid, not UN-approved
    Bucket with plastic handle and lid, not UN-approved
    White
    3,0 L
    198 mm
    172 mm
    Log in for availability
    Log in for availability
    1 * 120 ST
    1 * 120 ST - 758,00 €
    758,00 €
    1 * 120 ST
       758,00 €
  • RIXI7-1001-050
    Tamper proof bucket with plastic handle, not UN-approved
    Tamper proof bucket with plastic handle, not UN-approved
    White
    5,0 L
    224 mm
    191,5 mm
    Log in for availability
    Log in for availability
    1 * 90 ST
    1 * 90 ST - 285,00 €
    285,00 €
    1 * 90 ST
       285,00 €
  • RIXI7-1001-050-K
    Tamper proof bucket with plastic handle, not UN-approved
    Tamper proof bucket with plastic handle, not UN-approved
    White
    5,0 L
    224 mm
    191,5 mm
    Log in for availability
    Log in for availability
    1 * 900 ST
    1 * 900 ST - 5 330,00 €
    5 330,00 €
    1 * 900 ST
       5 330,00 €
  • RIXI7-5101-055
    Bucket with plastic handle and lid, UN-Y approved
    Bucket with plastic handle and lid, UN-Y approved
    White
    5,5 L
    237 mm
    208 mm
    Log in for availability
    Log in for availability
    1 * 80 ST
    1 * 80 ST - 950,00 €
    950,00 €
    1 * 80 ST
       950,00 €
  • RIXIU-7S-1001-080K
    Tamper proof bucket with zinced steel handle and lid, not UN-approved
    Tamper proof bucket with zinced steel handle and lid, not UN-approved
    White
    8,0 L
    266 mm
    212 mm
    Log in for availability
    Log in for availability
    1 * 45 ST
    1 * 45 ST - 478,00 €
    478,00 €
    1 * 45 ST
       478,00 €
  • RIXI7-5101-110
    Bucket with plastic handle and lid, UN-Y approved
    Bucket with plastic handle and lid, UN-Y approved
    White
    11,0 L
    293 mm
    245 mm
    Log in for availability
    Log in for availability
    1 * 40 ST
    1 * 40 ST - 614,00 €
    614,00 €
    1 * 40 ST
       614,00 €
  • RIXI7-1001-100-K
    Tamper proof bucket with plastic handle, not UN-approved (without lid, must be ordered separately)
    Tamper proof bucket with plastic handle, not UN-approved (without lid, must be ordered separately)
    White
    10,0 L
    266 mm
    262,5 mm
    Log in for availability
    Log in for availability
    1 * 480 ST
    1 * 480 ST - 5 000,00 €
    5 000,00 €
    1 * 480 ST
       5 000,00 €
  • RIXI7-1001-100
    Tamper proof bucket with plastic handle, not UN-approved (without lid, must be ordered separately)
    Tamper proof bucket with plastic handle, not UN-approved (without lid, must be ordered separately)
    White
    10,0 L
    266 mm
    262,5 mm
    Log in for availability
    Log in for availability
    1 * 48 ST
    1 * 48 ST - 354,00 €
    354,00 €
    1 * 48 ST
       354,00 €
  • RIXI7-5101-180
    Bucket with plastic handle and lid, UN-X approved
    Bucket with plastic handle and lid, UN-X approved
    White
    18,0 L
    334 mm
    290 mm
    Log in for availability
    Log in for availability
    1 * 180 ST
    1 * 180 ST - 4 790,00 €
    4 790,00 €
    1 * 180 ST
       4 790,00 €
  • RIXI7-2001-180
    Bucket with carrying handle and handle roller, not UN-approved
    Bucket with carrying handle and handle roller, not UN-approved
    White
    18,0 L
    298 mm
    298 mm
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST - 392,00 €
    392,00 €
    1 * 30 ST
       392,00 €
  • RIXI7-1001-200
    Bucket with metal handle, not UN-approved (without lid, must be ordered seperately)
    Bucket with metal handle, not UN-approved (without lid, must be ordered seperately)
    White
    20,0 L
    326 mm
    327 mm
    Log in for availability
    Log in for availability
    1 * 15 ST
    1 * 15 ST - 265,00 €
    265,00 €
    1 * 15 ST
       265,00 €
  • RIXI7-1001-300
    Bucket with recessed grips, not UN-approved (without lid, must be ordered seperately)
    Bucket with recessed grips, not UN-approved (without lid, must be ordered seperately)
    White
    30,0 L
    385 mm
    388 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 241,00 €
    241,00 €
    1 * 10 ST
       241,00 €
  • RIXI7-1001-300-K
    Bucket with recessed grips, not UN-approved (without lid, must be ordered seperately)
    Bucket with recessed grips, not UN-approved (without lid, must be ordered seperately)
    White
    30,0 L
    385 mm
    388 mm
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 6 270,00 €
    6 270,00 €
    1 * 200 ST
       6 270,00 €
  • RIXI7-5101-600
    Bucket with recessed grips, not UN-approved (without lid, must be ordered separately)
    Bucket with recessed grips, not UN-approved (without lid, must be ordered separately)
    White
    60,0 L
    360 mm
    565 mm
    Log in for availability
    Log in for availability
    1 * 125 ST
    1 * 125 ST - 9 470,00 €
    9 470,00 €
    1 * 125 ST
       9 470,00 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Lever lid for 2,5 L bucket, PP, water proof
    Lever lid for 2,5 L bucket, PP, water proof
    RIXI8-7100-025
    Log in for availability
    1 * 1 ST - 1,70 €
Product Information
  • Catalog #
    Availability
    Price
  • Lever lid, for 5 L bucket, PP, water proof
    Lever lid, for 5 L bucket, PP, water proof
    RIXI8-7100-050
    Log in for availability
    1 * 1 ST - 2,10 €
Product Information
  • Catalog #
    Availability
    Price
  • Lever lid for 10 L bucket, PP, water proof
    Lever lid for 10 L bucket, PP, water proof
    RIXI8-7100-100
    Log in for availability
    1 * 48 ST - 102,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Lid for 15 l bucket, oval, PP
    Lid for 15 l bucket, oval, PP
    RIXI8-7200-180
    Log in for availability
    1 * 36 ST - 123,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Lever lid for 2 l bucket, PP, water proof
    Lever lid for 2 l bucket, PP, water proof
    RIXI8-7100-018
    Log in for availability
    1 * 180 ST - 259,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Lever lid for 1 l bucket, PP, water proof
    Lever lid for 1 l bucket, PP, water proof
    RIXI8-7100-010
    Log in for availability
    1 * 25 ST - 16,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Lever lid for 0,6 l bucket, PP, water proof
    Lever lid for 0,6 l bucket, PP, water proof
    RIXI8-7100-006
    Log in for availability
    1 * 435 ST - 262,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Lever lid for 20 l bucket, PP, water proof
    Lever lid for 20 l bucket, PP, water proof
    RIXI8-7100-210
    Log in for availability
    1 * 15 ST - 62,60 €
Product Information
  • Catalog #
    Availability
    Price
  • Lever lid for 30 l bucket, PP, water proof
    Lever lid for 30 l bucket, PP, water proof
    RIXI8-7100-300
    Log in for availability
    1 * 10 ST - 63,80 €