Orders LinkContactUsLinkComponent
Faecal containers with screw cap
Faecal containers with screw cap
Faecal containers with screw cap
Supplier:  LP ITALIANA
Certificates
Faecal containers with screw cap
Faecal containers with screw cap
8 Options Available Below
Product Details & Documents
Containers made of PP, translucent.
  • Excellent chemical resistance
  • Excellent temperature resistance
  • Easy to clean
With or without spoon and label.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Documents
Product Family Options
Product Information
  • Capacity
    Type
    Material
    Ext.Ø
    Height
    Availability
    Price
  • 25 ml
    without label and spoon
    PP
    25 mm
    80 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 232,00 €
    232,00 €
    1 * 1.000 ST
       232,00 €
  • 25 ml
    with label, without spoon
    PP
    25 mm
    80 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 271,00 €
    271,00 €
    1 * 1.000 ST
       271,00 €
  • 25 ml
    with label and spoon
    PP
    25 mm
    80 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 246,00 €
    246,00 €
    1 * 1.000 ST
       246,00 €
  • 25 ml
    with label and spoon, sterile, individually wrapped
    PP
    25 mm
    80 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 296,00 €
    296,00 €
    1 * 1.000 ST
       296,00 €
  • 25 ml
    with spoon, without label
    PP
    25 mm
    80 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 237,00 €
    237,00 €
    1 * 1.000 ST
       237,00 €
  • 25 ml
    with spoon, without label
    PP
    25 mm
    80 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 227,00 €
    227,00 €
    1 * 1.000 ST
       227,00 €
  • 25 ml
    with label, without spoon, sterile
    PP
    25 mm
    80 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 251,00 €
    251,00 €
    1 * 1.000 ST
       251,00 €
  • 25 ml
    without label and spoon, sterile
    PP
    25 mm
    80 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 237,00 €
    237,00 €
    1 * 1.000 ST
       237,00 €
This product has been blocked by your organization. Please contact your purchasing department for more information