Orders LinkContactUsLinkComponent
Adapters, bends
Adapters, bends
29/32 NS
Catalog # WITG1.008.007
Supplier:   witeg Labortechnik
Adapters, bends
Catalog # WITG1.008.007
Supplier:  witeg Labortechnik
Adapters, bends
Adapters, bends
29/32 NS
Price
28,60 €
Log in for availability
1 * 1 ST
Product Details & Documents
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Inclined at an angle of 105°. With 2 standard ground cones.
Specifications
  • Cone size:
    29/32 NS