Orders LinkContactUsLinkComponent
Bends
Bends
19/26 + 24/29 NS
Catalog # WITG1.000.004
Supplier:   witeg Labortechnik
Bends
Catalog # WITG1.000.004
Supplier:  witeg Labortechnik
Bends
Bends
19/26 + 24/29 NS
Price
31,70 €
Log in for availability
1 * 1 ST
Product Details & Documents
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Inclined at an angle of 90°. With 2 standard ground cones.
Specifications
  • Cone size:
    19/26 + 24/29 NS