ContactUsLinkComponent
Adapter-S, KF, for reaction vessels with cooling jacket
Adapter-S, KF, for reaction vessels with cooling jacket
Catalog # WITG0.998.129
Supplier:  witeg Labortechnik
undefined
Adapter-S, KF, for reaction vessels with cooling jacket
Catalog # WITG0.998.129
Supplier:  witeg Labortechnik
Specifications
  • Socket:
    KF 29/15
  • Type:
    with hose connection 12 mm
  • Pk:
    1
Specifications
About this item
Borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
S-adapters for thermostatic reaction vessels with cooling jacket.