Screws with cylindrical head, BOLA
Supplier: Bohlender
Certificates
About this item
PTFE, white.
- Temperature resistance: from -200 °C to +250 °C
- Universal chemical resistance
- FDA conform
- Similar to DIN 84/DIN EN ISO 1207
Frequently Bought Together