Order Entry
Austria
ContactUsLinkComponent
Infusion filter, Sterifix®
Infusion filter, Sterifix®
Catalog # BRAU4099303
Supplier:  B. Braun
undefined
Infusion filter, Sterifix®
Catalog # BRAU4099303
Supplier:  B. Braun

Specifications

  • Description:
    Infusion filters, Sterifix®
  • Pk:
    50

Specifications

About this item

Sterifix infusion filter is specially designed for bacteria and particle retention.

  • Reliable air venting regardless of position
  • High throughput 0,2 μm Supor® membrane
  • Pressure resistant to 3,1 bar Luer lock fitting
  • Low absorption rate of proteins