Order Entry
Austria
ContactUsLinkComponent
Sterilin™ Petri Dishes
Sterilin™ Petri Dishes
Catalog # SLIN502VF
Supplier:  Sterilin
undefined
Sterilin™ Petri Dishes
Catalog # SLIN502VF
Supplier:  Sterilin

Specifications

  • Ø:
    90 mm
  • Sterility:
    Aseptic
  • Version:
    With 2 compartments, 3 vents
  • Description:
    Petri dish, 2 compartment, 90 mm, clear colour, round shape
  • Height:
    16 mm
  • Pk:
    500

Specifications

About this item

Ideal for use with different media or when savings in media or incubator space are required. Perform nonstandard applications with Thermo Scientific™ Sterilin™ Special Petri Dishes. Minimum order quantities apply. Not available in America.

  • Aseptically manufactured under clean room conditions (class 7 ISO14644) to exclude microbiological contamination
  • Single frosted marking area on the base of the dish - ideal for plate orientation with automatic pouring systems
  • CE marked in accordance with European Directive 98/79/EC
  • Manufactured to BS 611 part 2
  • In vitro use only