Order Entry
Austria
ContactUsLinkComponent
Sudan black B
Sudan black B
Catalog # ROTH0292.1
Supplier:  Roth Carl
CAS Number:  
Sudan black B
Catalog # ROTH0292.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236
  • Environmentally Preferable:

Specifications

About this item

Synonyms: CI 26150, Ceres black BN, Solvent Black 3, Fat Black HB
 :
 :456.55 g/mol
Storage Temperature:Ambient
EINECS:224-087-1
CAS Number:4197-25-5
Formula:C₂₉H₂₄N₆
MDL Number:MFCD00006919