5-Iodo-1,3-dimethyluracil, Sigma-Aldrich®
Catalog # SIAL415294-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:266.04 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:40738-83-8
Formula:C₆H₇IN₂O₂
MDL Number:MFCD00192228