
HEPPSO (β-hydroxy-4-(2-hydroxyethyl)piperazine-1-propanesulphonic acid)
Artikelnr. A1072.0100
Supplier: PanReac AppliChem
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:268.3305 g/mol
EINECS:269-990-1
:GHS08
Storage Temperature:Ambient
CAS Number:68399-78-0
Formula:C₉H₂₀N₂O₅S
MDL Number:MFCD00038351