Order Entry
Austria
ContactUsLinkComponent
Amphotericin B 250 µg/ml in aqueous solution, HyClone™
Amphotericin B 250 µg/ml in aqueous solution, HyClone™
Catalog # HYCLSV30078.01
CAS Number:  
undefined
Amphotericin B 250 µg/ml in aqueous solution, HyClone™
Catalog # HYCLSV30078.01
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 mL
  • Seq:
    2370

Specifications

Specification Test Results

  • Sterilisation:
    0,2 µm sterile filtered
  • Antibiotic Spectrum:
    Antimycotic, Antifungal
  • Concentration:
    250 µg/ml

Specification Test Results

About this item

 :
 :924.08 g/mol
Storage Temperature:Freezer
EINECS:215-742-2
CAS Number:1397-89-3
Formula:C₄₇H₇₃NO₁₇
MDL Number:MFCD00877763