Order Entry
Austria
ContactUsLinkComponent
Acriflavine fluorescent labeling reagent, Sigma-Aldrich®
Acriflavine fluorescent labeling reagent, Sigma-Aldrich®
Certificates
undefined
Acriflavine fluorescent labeling reagent, Sigma-Aldrich®

Specification Test Results

  • Appearance (Form):
    Powder
  • Proton NMR spectrum:
    Conforms to Structure
  • EmM, 462 nm (+/- 3 nm), C = 0.004 g/L in Methanol:
    ≥48000
  • Wavelength:
    459 - 465 nm
  • EmM, 262 nm (+/- 3 nm), C = 0.004 g/L in Methanol:
    ≥50000
  • Solubility (Colour):
    Orange to Brown
  • Solubility (Turbidity), 50 mg/mL in H₂O:
    Clear

Specification Test Results

Frequently Bought Together