Acriflavine neutral 13.3-15.8% available chlorine
:8048-52-0
:65589-70-0
:C₂₇H₂₅ClN₆
:MFCD00064307
:468.9806 g/mol
:179…181 °C
:GHS07
:Ambient
- Appearance:Orange to red-brown Powder
- Infrared spectrum:Conforms
- Titration Argentometric:13.3 to 15.8 % (Chlorine) on dried substance
- Loss on drying:≤8 % (105°C, 2 h)
Frequently Bought Together