Order Entry
Austria
ContactUsLinkComponent
Octyl-β-D-thioglucopyranoside, Calbiochem®
Octyl-β-D-thioglucopyranoside, Calbiochem®
Catalog # 494461-5
CAS Number:  
Octyl-β-D-thioglucopyranoside, Calbiochem®
Catalog # 494461-5
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :308.44 g/mol
Storage Temperature:Freezer
CAS Number:85618-21-9
Formula:C₁₄H₂₈O₅S
MDL Number:MFCD00012189