Order Entry
Austria
ContactUsLinkComponent
Soy bean trypsin inhibitor
Soy bean trypsin inhibitor
Certificates
Soy bean trypsin inhibitor

Specifications

  • CAS No.:
    9035-81-8
  • Species:
    Soy bean
  • Storage conditions:
    Freezer
  • Protein/peptide name:
    Trypsin inhibitor

Specifications

Frequently Bought Together