L-Lactate dehydrogenase Typ II (from Muscle)
Supplier: Merck
Certificates
About this item
Specifications
- CAS No.:9001-60-9
- Source:Muscle
- Storage conditions:Refrigerator
- MDL No.:MFCD00131466
- Enzyme name:L-Lactate dehydrogenase, Typ II
- Enzyme synonyms:LDH
Specifications
Frequently Bought Together