Order Entry
Austria
ContactUsLinkComponent
L-Lactate dehydrogenase Typ XI (from Muscle)
L-Lactate dehydrogenase Typ XI (from Muscle)
Supplier:  Merck
Certificates
L-Lactate dehydrogenase Typ XI (from Muscle)
Supplier:  Merck

Specifications

  • CAS No.:
    9001-60-9
  • Source:
    Muscle
  • Storage conditions:
    Refrigerator
  • MDL No.:
    MFCD00131466
  • Enzyme name:
    L-Lactate dehydrogenase, Typ XI
  • Enzyme synonyms:
    LDH

Specifications

Frequently Bought Together