Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Enzymes
Pig L-Lactate dehydrogenase (from Heart)
Pig L-Lactate dehydrogenase (from Heart)
Pig L-Lactate dehydrogenase (from Heart)
427211-50
:
Merck Millipore (Calbiochem)
:
Pig L-Lactate dehydrogenase (from Heart)
427211-50
:
Merck Millipore (Calbiochem)
:
Pk:
50 KU
CAS No.:
9001-60-9
Source:
Heart
Species:
Pig
Storage conditions:
Refrigerator
MDL No.:
MFCD00131466
Enzyme name:
L-Lactate dehydrogenase
Enzyme synonyms:
LDH