Vancomycin hydrochloride
Catalog # MOLE16685638-1G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:1485.71 g/mol
CAS Number:1404-93-9
:123409-00-7
Formula:C₆₆H₇₅Cl₂N₉O₂₄·HCl
MDL Number:MFCD03613611