VWR®, Pepstatin A
Catalog # M183-5MG
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 mg
- CAS No.:26305-03-3
- Storage conditions:Refrigerator
- MDL No.:MFCD00060740
- Protein/peptide name:Pepstatin A
- Grade:Proteomics Grade
- Molecular formula:C₃₄H₆₃N₅O₉
Specifications
About this item
An aspartic protease inhibitor of pepsin, renin, and cathepsin D. Can also block HIV protease. Not soluble in water.
FTIR: Suitable
Melting point: 220-240 ° C
Optical rotation (0.5%, MeOH): -95 to -85 °
Purity (HPLC): 75%