Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Acetyl-Pepstatin
Acetyl-Pepstatin
Acetyl-Pepstatin
Catalog # 110175-1
Supplier:
Merck Millipore (Calbiochem)
CAS Number:
Expand image
Acetyl-Pepstatin
Catalog # 110175-1
Supplier:
Merck Millipore (Calbiochem)
CAS Number:
Specifications
Pack type:
Glass bottle for solids
Pk:
1 mg
CAS No.:
28575-34-0
Protein/peptide name:
Acetyl-Pepstatin
Specifications
About this item
Product Details
Product Details