VWR®, Leupeptin hemisulphate
Catalog # M180-5MG
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 mg
- CAS No.:103476-89-7
- MDL No.:MFCD00037012
- Protein synonyms:(Ac-Leu-Leu-Arg-al hemisulphate) monohydrate
- Protein/peptide name:Leupeptin hemisulphate
- Grade:Proteomics Grade
- Molecular formula:C₄₀H₇₈N₁₂O₁₂S
Specifications
About this item
A reversible inhibitor of cysteine proteases and serine proteases resembling trypsin. Inhibits trypsin, plasmin, papain, kallikrein, thrombin, and cathepsin A and B.
- Product is animal-free
Extracted proteins are exposed to the threat of degradation by proteases from the moment cells or tissues are lysed. Proteases present in the living cell are released from their regulated environment into the extract where they may freely degrade their targets, resulting in drastically reduced protein yield and sample quality. Complex protein solutions are typically treated with a mixture of several protease inhibitors in order to preserve the native state of the proteins.