VWR®, Leupeptin hemisulphate
A reversible inhibitor of cysteine proteases and serine proteases resembling trypsin. Inhibits trypsin, plasmin, papain, kallikrein, thrombin, and cathepsin A and B.
- CAS No.:103476-89-7
- Protein synonyms:(Ac-Leu-Leu-Arg-al hemisulphate) monohydrate
- Protein/peptide name:Leupeptin hemisulphate
- Grade:Ultra Pure Grade
- Molecular formula:C₄₀H₇₈N₁₂O₁₂S
Frequently Bought Together