Order Entry
Austria
Orders LinkContactUsLinkComponent
Actinomycin D

Actinomycin D

Glass bottle for solids, 5 mg
Catalog # ROTH8969.1
Marketsource
Supplier:   Roth Carl
Actinomycin D
Catalog # ROTH8969.1
Supplier:  Roth Carl
Marketsource
CAS Number:  50-76-0
Actinomycin D

Glass bottle for solids, 5 mg

CAS Number:  50-76-0
Price
134,00 €
Log in for availability
1 * 5 mg
Product Details & Documents
  • CAS Number:50-76-0
  • Formula:C₆₂H₈₆N₁₂O₁₆
  • MDL Number:MFCD00005033
  • Molecular Weight:1255.5 g/mol
  • UN:3462
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.