Phosphoramidon (disodium salt)
Catalog # 525276-5
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 mg
- CAS No.:119942-99-3
- Storage conditions:Freezer
- Protein/peptide name:Phosphoramidon (disodium salt)