

Thymidine (2'-deoxythymidine), Millipore®
Catalog # 6060-5
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:242.23 g/mol
EINECS:200-070-4
:GHS08
:GHS07
Storage Temperature:Ambient
CAS Number:50-89-5
:146183-25-7
Formula:C₁₀H₁₄N₂O₅
MDL Number:MFCD00006537