Order Entry
Austria
ContactUsLinkComponent
Adenosine-5'-triphosphate disodium salt (ATP disodium salt)
Adenosine-5'-triphosphate disodium salt (ATP disodium salt)
Catalog # MOLE16368689-25G
Supplier:  Molekula
CAS Number:  
Adenosine-5'-triphosphate disodium salt (ATP disodium salt)
Catalog # MOLE16368689-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308
  • Environmentally Preferable:

Specifications

About this item

Synonyms: ((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl tetrahydrogen triphosphate, disodium salt, Adenosine 5'-triphosphoric acid disodium, Adenosine 5′-triphosphate disodium salt, ATP 2Na, ATP disodium salt, Adenosine 5ʹ-Triphosphate, Disodium Salt
 :
 :551.14 g/mol
EINECS:213-579-1
CAS Number:987-65-5
 :MFCD00080339
Formula:C₁₀H₁₄N₅Na₂O₁₃P₃