Adenosine 5'-monophosphate monohydrate (from yeast) ≥97%, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:18422-05-4
:MFCD00005750
Formula:C₁₀H₁₄N₅O₇P·1H₂O
MDL Number:MFCD00149360
:365.24 g/mol
EINECS:200-500-0
Frequently Bought Together