Order Entry
Austria
ContactUsLinkComponent
Adenosine 5'-monophosphate monohydrate
Adenosine 5'-monophosphate monohydrate
Catalog # MOLEM81117744
Supplier:  Molekula
CAS Number:  
Adenosine 5'-monophosphate monohydrate
Catalog # MOLEM81117744
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :365.24 g/mol
EINECS:200-500-0
CAS Number:18422-05-4
 :MFCD00005750
Formula:C₁₀H₁₄N₅O₇P·1H₂O
MDL Number:MFCD00149360