Adenosine 5'-diphosphate disodium salt (ADP disodium salt)
Catalog # MOLE22770490-1G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:471.167 g/mol
EINECS:240-314-7
CAS Number:16178-48-6
Formula:C₁₀H₁₃N₅Na₂O₁₀P₂
MDL Number:MFCD00066635