2'-Deoxyadenosine 5'-triphosphate disodium salt (dATP disodium salt)
Catalog # MOLEM61986848
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:535.14 g/mol
EINECS:277-809-2
CAS Number:74299-50-6
Formula:C₁₀H₁₄N₅Na₂O₁₂P₃
MDL Number:MFCD00080335