Order Entry
Austria
ContactUsLinkComponent
2',5'-Dideoxyadenosine, Millipore®
2',5'-Dideoxyadenosine, Millipore®
Catalog # 288104-1
CAS Number:  
2',5'-Dideoxyadenosine, Millipore®
Catalog # 288104-1
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :235.2425 g/mol
CAS Number:6698-26-6
Formula:C₁₀H₁₃N₅O₂
MDL Number:MFCD00210904