2'-Deoxyguanosine-5'-triphosphate trisodium salt (dGTP-Na3), Sigma-Aldrich®
Catalog # SIALD7170-.5ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:0,5 mL
- Seq:2040
Specifications
About this item
:
:573.12 g/mol
EINECS:300-026-5
CAS Number:93919-41-6
Formula:C₁₀H₁₃N₅Na₃O₁₃P₃
MDL Number:MFCD00080337