Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tobramycin
Tobramycin
Tobramycin
Tobramycin
EHERCA17588000
:
EHRENSTORFER
:
Tobramycin
EHERCA17588000
:
EHRENSTORFER
:
Pack type:
Glass bottle for solids
Pk:
0,1 g
Seq:
3020
:
:
467.51 g/mol
:
251-322-5
:
GHS08
:
GHS07
:
Refrigerator
:
32986-56-4
:
C₁₈H₃₇N₅O₉
:
MFCD00077885