Acridine orange base ≥75% (dye content), Sigma-Aldrich®
:494-38-2
:
:C₁₇H₁₉N₃
:MFCD00005029
:265.35 g/mol
:165 °C (dec., lit.)
:M-Clarity™ quality level = MQ200
:Ambient
- Appearance (Form):Powder
- Infrared spectrum:Conforms to Structure
- Wavelength:486 - 492 nm
- Extinction Coefficient:≥35000
- Carbon:≥56.2 %
- Nitrogen:≥11.6 %
- Purity (HPLC):≥75 %
- Solubility (Turbidity):Clear
- Solubility (Color); Orange to Very Dark Red-Brown, c = 1mg/ml, 1N HCl:Conforms to Requirements
Frequently Bought Together