Sudan red 7B (fat red bluish, solvent red 19, fat red 7B), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:6368-72-5
:
Formula:C₂₄H₂₁N₅
MDL Number:MFCD00003904
:379.4570 g/mol
Melting Pt:130 °C (dec., lit.)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:228-862-5
Specification Test Results
- Appearance (color):Red to very dark red and purple to very dark purple and brown to very dark brown and very dark red-brown
- Appearance (form):Powder
- Thin layer chromatogr.:Consistent with listing
- Solubility (color):Red to very dark red
- Solubility (turbidity):Clear to turbid
- Solubility (method):10mg/ml (1%), acetone
- Carbon content:72 - 77 %
- Nitrogen content:17 - 19 %
- Infrared spectrum:Conforms to structure
- Wavelength (1) (uv):528 - 538 nm
- Molar absorbancy index (1):≥ 24660
- Wavelength (2) (uv):367 - 377 nm
- Molar absorbancy index (2):≥ 14230
- Solvent (uv):Toluene
- Recommended retest period:48 months
Specification Test Results
Frequently Bought Together