Order Entry
Austria
ContactUsLinkComponent
Sudan red 7B (fat red bluish, solvent red 19, fat red 7B), Supelco®
Sudan red 7B (fat red bluish, solvent red 19, fat red 7B), Supelco®
Catalog # 53373-25MG
Supplier:  Merck
CAS Number:  
undefined
Sudan red 7B (fat red bluish, solvent red 19, fat red 7B), Supelco®
Catalog # 53373-25MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :379.4570 g/mol
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:228-862-5
CAS Number:6368-72-5
Formula:C₂₄H₂₁N₅
MDL Number:MFCD00003904