Sudan red 7B (fat red bluish, solvent red 19, fat red 7B), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:6368-72-5
:
Formula:C₂₄H₂₁N₅
MDL Number:MFCD00003904
:379.4570 g/mol
Storage Temperature:Ambient
EINECS:228-862-5
Frequently Bought Together