Order Entry
Austria
ContactUsLinkComponent
Sudan red 7B (fat red bluish, solvent red 19, fat red 7B)
Sudan red 7B (fat red bluish, solvent red 19, fat red 7B)
Catalog # EHERC16986120
Supplier:  EHRENSTORFER
CAS Number:  
Sudan red 7B (fat red bluish, solvent red 19, fat red 7B)
Catalog # EHERC16986120
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
 :379.4570 g/mol
Storage Temperature:Ambient
EINECS:228-862-5
CAS Number:6368-72-5
Formula:C₂₄H₂₁N₅
MDL Number:MFCD00003904