
Ponceau S
Catalog # A2935.0500
Supplier: PanReac AppliChem
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
Specifications
About this item
:
:760.56 g/mol
EINECS:228-319-2
:GHS07
Storage Temperature:Ambient
CAS Number:6226-79-5
Formula:C₂₂H₁₂N₄Na₄O₁₃S₄
MDL Number:MFCD00003892