Order Entry
Austria
ContactUsLinkComponent
Ponceau S, Proteomics Grade
Irritant
Ponceau S, Proteomics Grade
Catalog # K793-500ML
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Ponceau S, Proteomics Grade
Catalog # K793-500ML
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Abs.@ Lambda Max (1:50, Water):
    0.9 - 1.4
  • Lambda Max (1:50, Water):
    517 nm - 523 nm

Specification Test Results

About this item

 :
 :760.56 g/mol
EINECS:228-319-2
 :GHS07
Storage Temperature:Ambient
CAS Number:6226-79-5
Formula:C₂₂H₁₂N₄Na₄O₁₃S₄
MDL Number:MFCD00003892