Order Entry
Austria
ContactUsLinkComponent
3,3'-Dichlorobenzidine
Catalog # EHERC12377900
Supplier:  EHRENSTORFER
CAS Number:  
3,3'-Dichlorobenzidine
Catalog # EHERC12377900
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
EINECS:202-109-0
UN:2811
ADR:6.1,III
CAS Number:91-94-1
Formula:C₁₂H₁₀Cl₂N₂